ENAH11E41589 Enamine

2,2-difluorocyclooctan-1-amine hydrochloride

  • MDL number MFCD31666527
  • Cas Number : 75149-51-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008132

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008132
Availability details for
Quantity
  • Availability
  • Add to Cart