ENAH11E4158B Enamine

1-(4-chlorophenyl)-1-(2-fluorophenyl)methanamine

  • MDL number MFCD09810604
  • Cas Number : 1016748-07-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008137

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008137
Availability details for
Quantity
  • Availability
  • Add to Cart