ENAH11E4158C Enamine

1-(4-chlorophenyl)-1-[3-(trifluoromethyl)phenyl]methanamine

  • MDL number MFCD11574791
  • Cas Number : 735209-67-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008138

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008138
Availability details for
Quantity
  • Availability
  • Add to Cart