ENAH11E4159A Enamine

4-[(1E)-1-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)prop-1-en-2-yl]pyridine

  • MDL number MFCD31666596
  • Cas Number : 1331936-58-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008181

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008181
Availability details for
Quantity
  • Availability
  • Add to Cart