ENAH11E4159E Enamine

N-(cyclopropylmethyl)pyrrolidine-3-sulfonamide hydrochloride

  • MDL number MFCD31666849
  • Cas Number : 2228694-22-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008215

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008215
Availability details for
Quantity
  • Availability
  • Add to Cart