ENAH11E415A5 Enamine

1-(3,4-dichlorophenyl)-3-phenylpropan-1-amine hydrochloride

  • MDL number MFCD31666678
  • Cas Number : 2229135-08-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008231

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008231
Availability details for
Quantity
  • Availability
  • Add to Cart