ENAH11E415B6 Enamine

methyl 3-chloro-2,2-dimethyl-3-oxopropanoate

  • MDL number MFCD28517441
  • Cas Number : 64507-20-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008419

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008419
Availability details for
Quantity
  • Availability
  • Add to Cart