ENAH11E415B7 Enamine

ethyl 7-hydroxy-[1,2,4]triazolo[1,5-a]pyrimidine-6-carboxylate

  • MDL number MFCD00086159
  • Cas Number : 52893-00-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008421

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008421
Availability details for
Quantity
  • Availability
  • Add to Cart