ENAH11E415BD Enamine

1-(4-chlorophenyl)-1-(2-methoxyphenyl)methanamine

  • MDL number MFCD09804539
  • Cas Number : 1016696-64-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008449

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008449
Availability details for
Quantity
  • Availability
  • Add to Cart