ENAH11E415BF Enamine

1-{7,7-difluorobicyclo[4.1.0]heptan-3-yl}methanamine hydrochloride

  • MDL number MFCD31666614
  • Cas Number : 2228326-01-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008459

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008459
Availability details for
Quantity
  • Availability
  • Add to Cart