ENAH11E415C1 Enamine

2-{4-[(tert-butoxy)carbonyl]-4-azaspiro[2.5]octan-7-yl}acetic acid

  • MDL number MFCD31666535
  • Cas Number : 2227206-39-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008463

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008463
Availability details for
Quantity
  • Availability
  • Add to Cart