ENAH11E415C4 Enamine

2-hydroxybicyclo[3.3.1]nonan-9-one

  • MDL number MFCD24689236
  • Cas Number : 2568-21-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008470

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008470
Availability details for
Quantity
  • Availability
  • Add to Cart