ENAH11E415CC Enamine

4-amino-N-(1,3-dioxaindan-5-yl)benzamide

  • MDL number MFCD10021679
  • Cas Number : 1831221-88-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008510

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008510
Availability details for
Quantity
  • Availability
  • Add to Cart