ENAH11E415D3 Enamine

2-methyl-6-[3-(trifluoromethyl)-1,2,4-oxadiazol-5-yl]phenol

  • MDL number MFCD25257958
  • Cas Number : 1550662-56-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008636

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008636
Availability details for
Quantity
  • Availability
  • Add to Cart