ENAH11E415D7 Enamine

2-bromo-5,5-dimethyl-4,5,6,7-tetrahydro-1-benzofuran-4-one

  • MDL number MFCD31666562
  • Cas Number : 2092225-00-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008648

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008648
Availability details for
Quantity
  • Availability
  • Add to Cart