ENAH11E415D8 Enamine

potassium 3-[3-(tert-butoxy)-3-oxopropyl]pyridine-2-carboxylate

  • MDL number MFCD31666563
  • Cas Number : 2229558-05-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data K+
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008651

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008651
Availability details for
Quantity
  • Availability
  • Add to Cart