ENAH11E415D9 Enamine

4'-(aminomethyl)-[1,1'-biphenyl]-3-carboxylic acid hydrochloride

  • MDL number MFCD31666517
  • Cas Number : 578026-45-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008652

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008652
Availability details for
Quantity
  • Availability
  • Add to Cart