ENAH11E415E4 Enamine

N-(adamantan-1-yl)-N'-ethylguanidine hydrochloride

  • Cas Number : 23166-32-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008827

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008827
Availability details for
Quantity
  • Availability
  • Add to Cart