ENAH11E415EA Enamine

3-amino-1-methylcyclopentane-1-carboxylic acid hydrochloride

  • MDL number MFCD31666625
  • Cas Number : 2109088-29-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008842

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008842
Availability details for
Quantity
  • Availability
  • Add to Cart