ENAH11E415F1 Enamine

1-ethenyl-1H-1,2,3-triazole

  • MDL number MFCD31666569
  • Cas Number : 41917-22-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008887

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008887
Availability details for
Quantity
  • Availability
  • Add to Cart