ENAH11E415F3 Enamine

{2-azabicyclo[2.2.2]octan-1-yl}methanol hydrochloride

  • MDL number MFCD28166342
  • Cas Number : 1630906-36-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008938

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008938
Availability details for
Quantity
  • Availability
  • Add to Cart