ENAH2C555E01 Enamine

4-oxocycloheptane-1-carboxylic acid

  • MDL number MFCD11053707
  • Cas Number : 90482-29-4
  • Molecular Weight : 156.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-106834

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106834
Availability details for
Quantity
  • Availability
  • Add to Cart