ENAH2C555E35 Enamine

1,4-diethyl 2-formylbutanedioate

  • MDL number MFCD00056287
  • Cas Number : 5472-38-8
  • Molecular Weight : 202.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-84972

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84972
Availability details for
Quantity
  • Availability
  • Add to Cart