ENAH2C5566F4 Enamine

3-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-5,6,7,8-tetrahydronaphthalene-2-carboxylic acid

  • MDL number MFCD31924015
  • Cas Number : 2243513-09-7
  • Molecular Weight : 413.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-650641

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-650641
Availability details for
Quantity
  • Availability
  • Add to Cart