ENAH2C556861 Enamine

ethyl 5-phenyl-1,3-thiazole-2-carboxylate

  • MDL number MFCD11976519
  • Cas Number : 58333-72-5
  • Molecular Weight : 233.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1212471

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1212471
Availability details for
Quantity
  • Availability
  • Add to Cart