ENAH2C557034 Enamine

4-(1,1,2-trifluoroethyl)aniline hydrochloride

  • MDL number MFCD31667886
  • Cas Number : 2241129-95-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1721393

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1721393
Availability details for
Quantity
  • Availability
  • Add to Cart