ENAH2C557036 Enamine

5-[(tert-butoxy)carbonyl]-4H,5H,6H,7H-pyrazolo[1,5-a]pyrazine-3-sulfinic acid

  • MDL number MFCD31667872
  • Cas Number : 2230803-48-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1721486

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1721486
Availability details for
Quantity
  • Availability
  • Add to Cart