ENAH2C55703F Enamine

2-{1-[(tert-butoxy)carbonyl]piperidin-4-yl}cyclopentane-1-carboxylic acid

  • MDL number MFCD31665368
  • Cas Number : 2241142-07-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1722421

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1722421
Availability details for
Quantity
  • Availability
  • Add to Cart