ENAH2C557040 Enamine

tert-butyl 3-[4-(aminomethyl)phenyl]-2,2-dimethylpropanoate

  • MDL number MFCD31667606
  • Cas Number : 2230803-87-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1722500

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1722500
Availability details for
Quantity
  • Availability
  • Add to Cart