ENAH2C557042 Enamine

ethyl 3-(prop-2-yn-1-yl)pyrrolidine-3-carboxylate

  • MDL number MFCD31668787
  • Cas Number : 2243508-12-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1722538

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1722538
Availability details for
Quantity
  • Availability
  • Add to Cart