ENAH2C557045 Enamine

3-(methylamino)-7-oxa-1lambda6-thiaspiro[3.5]nonane-1,1-dione

  • MDL number MFCD31667878
  • Cas Number : 2230804-10-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1722563

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1722563
Availability details for
Quantity
  • Availability
  • Add to Cart