ENAH2C557052 Enamine

3-[(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)methyl]pyrrolidine hydrochloride

  • MDL number MFCD31668131
  • Cas Number : 2365173-93-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1722954

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1722954
Availability details for
Quantity
  • Availability
  • Add to Cart