ENAH2C557058 Enamine

(furan-2-yl)(imino)methyl-lambda6-sulfanone

  • MDL number MFCD31691279
  • Cas Number : 2230808-46-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1722978

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1722978
Availability details for
Quantity
  • Availability
  • Add to Cart