ENAH2C55705A Enamine

2-[(tert-butoxy)carbonyl]-7,7-difluoro-2-azabicyclo[4.1.0]heptane-5-carboxylic acid

  • MDL number MFCD31690027
  • Cas Number : 2241138-67-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1723097

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1723097
Availability details for
Quantity
  • Availability
  • Add to Cart