ENAH2C55705C Enamine

tert-butyl N-({5-oxospiro[3.5]nonan-6-yl}methyl)carbamate

  • MDL number MFCD31665372
  • Cas Number : 2241139-53-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1723441

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1723441
Availability details for
Quantity
  • Availability
  • Add to Cart