ENAH2C557061 Enamine

5-[(tert-butoxy)carbonyl]-6,6-dimethyl-1H,4H,5H,6H-pyrrolo[3,4-c]pyrazole-3-carboxylic acid

  • MDL number MFCD31668136
  • Cas Number : 2243511-99-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1723627

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1723627
Availability details for
Quantity
  • Availability
  • Add to Cart