ENAH2C557066 Enamine

tert-butyl N-{2-[4-(chlorosulfonyl)phenyl]ethyl}carbamate

  • MDL number MFCD31666925
  • Cas Number : 2230799-25-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1723802

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1723802
Availability details for
Quantity
  • Availability
  • Add to Cart