ENAH2C557068 Enamine

tert-butyl N-[2-(3-oxocyclopentyl)ethyl]carbamate

  • MDL number MFCD31668793
  • Cas Number : 2227154-95-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1723936

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1723936
Availability details for
Quantity
  • Availability
  • Add to Cart