ENAH2C557069 Enamine

(1Z)-buta-1,3-diene-1-sulfonyl fluoride

  • MDL number MFCD31667668

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1723962

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1723962
Availability details for
Quantity
  • Availability
  • Add to Cart