ENAH2C557075 Enamine

5-amino-1,3-dimethyl-1H-pyrazole-4-sulfonyl fluoride

  • MDL number MFCD31665637
  • Cas Number : 2241131-00-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1725578

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1725578
Availability details for
Quantity
  • Availability
  • Add to Cart