ENAH2C55707A Enamine

tert-butyl N-{[2-(hydroxymethyl)thiophen-3-yl]methyl}carbamate

  • MDL number MFCD31690950
  • Cas Number : 2230803-12-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1725952

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1725952
Availability details for
Quantity
  • Availability
  • Add to Cart