ENAH2C55707C Enamine

3-(2-aminoethyl)-4-chloroaniline

  • MDL number MFCD31668194
  • Cas Number : 2243514-27-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1726025

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1726025
Availability details for
Quantity
  • Availability
  • Add to Cart