ENAH2C557080 Enamine

1-[(iodomethyl)sulfanyl]ethan-1-one

  • MDL number MFCD31690953
  • Cas Number : 114254-42-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1726165

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1726165
Availability details for
Quantity
  • Availability
  • Add to Cart