ENAH2C557082 Enamine

2-tert-butyl-1-oxo-3-phenyl-octahydro-1H-isoindole-3a-carboxylic acid

  • MDL number MFCD31665639
  • Cas Number : 2241131-06-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1726201

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1726201
Availability details for
Quantity
  • Availability
  • Add to Cart