ENAH2C557083 Enamine

4-[(tert-butoxy)carbonyl]-2-[(prop-2-en-1-yloxy)methyl]morpholine-2-carboxylic acid

  • MDL number MFCD31667007
  • Cas Number : 2230798-46-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1726247

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1726247
Availability details for
Quantity
  • Availability
  • Add to Cart