ENAH2C557085 Enamine

4-(2-bromo-3-chlorophenyl)-1,2-dimethyl-1H-pyrrole-3-carboxylic acid

  • MDL number MFCD31667892
  • Cas Number : 2241128-26-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1726404

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1726404
Availability details for
Quantity
  • Availability
  • Add to Cart