ENAH2C557089 Enamine

2,5-dibromo-4-methoxybenzene-1-sulfonamide

  • MDL number MFCD31667012
  • Cas Number : 2230800-25-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2007688

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2007688
Availability details for
Quantity
  • Availability
  • Add to Cart