ENAH2C55708B Enamine

4-oxo-2-(trifluoromethyl)cyclopentane-1-carboxylic acid

  • MDL number MFCD31666933
  • Cas Number : 2241140-04-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2007800

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2007800
Availability details for
Quantity
  • Availability
  • Add to Cart