ENAH2C557092 Enamine

methyl 1-(1-methylpiperidin-4-yl)-5-oxopyrrolidine-3-carboxylate

  • MDL number MFCD31667063
  • Cas Number : 2230804-33-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008179

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008179
Availability details for
Quantity
  • Availability
  • Add to Cart