ENAH2C557098 Enamine

2-(methoxycarbonyl)-4H-thieno[3,2-b]pyrrole-5-carboxylic acid

  • MDL number MFCD31690926
  • Cas Number : 2241140-01-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008492

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008492
Availability details for
Quantity
  • Availability
  • Add to Cart